Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ST14 Rabbit Polyclonal Antibody

Supplier: BioVision

3855-100
10004-820EA 252.35 USD InStock
10004-820
Anti-ST14 Rabbit Polyclonal Antibody
Antibodies
Matriptase or MT-SP1 is an epithelial-derived type II transmembrane serine protease that is highly expressd in human cancer-derived cell lines. Matriptase cleaves and activates protease activated receptor-2, pro-urokinase plasminogen activator and pro-hepatocyte growth factor. Matriptase is strongly inhibited by the hepatocyte growth factor activator inhibitor type 1 (HAI-1), which is a reactive site loop of Kunitz domain and also a transmembrane serine protease inhibitor.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ST14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty