Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-INSIG1 Rabbit Polyclonal Antibody

Supplier: BioVision

3854-100
10004-816EA 298.7 USD InStock
10004-816
Anti-INSIG1 Rabbit Polyclonal Antibody
Antibodies
Insulin-induced gene (InSig) localizes in the endoplasmic recticulum(ER) and is highly expressed in the liver and fibroblast cell lines. InSig1 and InSig2 play important roles in the regulation of cholesterol biosynthesis. Sterol induces InSig1 binding to the sterol-sensing domain of SREBP cleavage-activating protein (SCAP). Both InSig1 and Insig2 prevent the export of SCAP from the ER and thus, inhibit cholesterol synthesis by preventing the proteolytic cleavage of SREBPs by the Golgi enzymes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: INSIG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Hamster

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty