Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDLBP Rabbit Polyclonal Antibody

Supplier: BioVision

3853-100
10004-988EA 252.35 USD InStock
10004-988
Anti-HDLBP Rabbit Polyclonal Antibody
Antibodies
High density lipoprotein-binding protein (HDLBP) or Vigilin is a protein that plays a role in the regulation and delivery of fat to cells for energy and storage. HDLBP binds HDL molecules and may function in the elimination of excess cellular cholesterol.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HDLBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty