Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FCN1 Rabbit Polyclonal Antibody

Supplier: BioVision

3852-100
10004-984EA 252.35 USD InStock
10004-984
Anti-FCN1 Rabbit Polyclonal Antibody
Antibodies
Ficolin-1 also known as M-Ficolin is a pattern recognition molecule of the lectin complement activation pathway. It is expressed mostly in monocytes, nuetrophils and in type II alveolar epithelial cells. Ficolin-1 is detected on the surface of circulating monocytes but is not present in the plasma. Ficolin-1 has a 28 aa signal sequence, an N-terminal collagen domain and a C-terminal fibrinogen-like (FBG) domain that contain a calcium binding site and a potential N-glycosylation site.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Ficolin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty