Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteinase Inhibitor 6 Mouse Monoclonal Antibody [clone: 3A]

Supplier: BioVision

3849-100
10004-976EA 303.85 USD InStock
10004-976
Anti-Proteinase Inhibitor 6 Mouse Monoclonal Antibody [clone: 3A]
Antibodies
Proteinase inhibitor 6 (PI-6) is a 42 kDa cytoplasmic protein belonging to the serine proteinase inhibitor (serpin) super family and the ovalbumin-serpin super family. Sequence similarity sμggests that PI-6 most resembles the ovalbumin serpins . Highest levels of PI-6 are found in skeletal muscle. PI-6 is also found in placenta, cardiac muscle, lung, liver, kidney and pancreas.

Recommended Dilutions: 0

Type: Primary
Antigen: Proteinase Inhibitor 6
Clonality: Monoclonal
Clone: 3A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty