Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BRK Rabbit Polyclonal Antibody

Supplier: BioVision

3847-100
10004-970EA 252.35 USD InStock
10004-970
Anti-BRK Rabbit Polyclonal Antibody
Antibodies
Breast tumor kinase, Brk (also known as PTK6), is a member of intracellular tyrosine kinase family that is thoμght to be related to Src family of tumor related kinases. Brk is expressed in breast carcinomas but not in normal breast tissue. The presence of Brk in the nucleus appears to be linked to suppression of tumor progression and has been shown to undergo autophosphorylation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BRK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty