Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DGAT1 Rabbit Polyclonal Antibody

Supplier: BioVision

3845-100
10004-964EA 252.35 USD InStock
10004-964
Anti-DGAT1 Rabbit Polyclonal Antibody
Antibodies
Diacylglycerol acyltransferase is also known as DGAT1 is a microsomal enzyme. It is involved in the synthesis of fatty acids into triglycerides. DGAT utilizes diacylglycerol (DAG) and fatty acyl CoA as substrates to synthesize triacylglycerol. DGAT plays an important role in the metabolism of cellular diacylglycerol and is essential in the physiologic processes of higher eukaryotes that includes triacylglycerol metabolism such as intestinal fat absorption, lipoprotein assembly, adipose tissue formation, and lactation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DGAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty