Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DMP1 Rabbit Polyclonal Antibody

Supplier: BioVision

3844-100
10004-960EA 288.4 USD InStock
10004-960
Anti-DMP1 Rabbit Polyclonal Antibody
Antibodies
DMP-1 (Dentin matrix protein1) is a member of the integrin ligand N-linked glycoprotein family.It is mostly found in bone, tooth and hypertrophic cartilage. DMP-1 plays an important role in dentin and bone mineralization. DMP-1 has multiple serine, glutamic acid and aspartic acid residues, a GAG attachement site and RGD motif. DMP-1 is proteolytically proceesed into mulitple fragemtns in the extracellular matrix (ECM) of bone and dentin.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty