Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LDHA Goat Polyclonal Antibody

Supplier: BioVision

3842-500
10004-602EA 231.75 USD InStock
10004-602
Anti-LDHA Goat Polyclonal Antibody
Antibodies
Lactate dehydrogenase (LDH) contains two isoforms in mammals, LDH-M and LDH-H, which come together to form a homotetramer of 36 kDa subunits. LDH is often as a marker for tissue breakdown or cytotoxicity. LDH shows cytoplasm localization.

Recommended Dilutions: Western Blot: 1:500-2500

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty