Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MATN2 Rabbit Polyclonal Antibody

Supplier: BioVision

3840-100
10004-812EA 252.35 USD InStock
10004-812
Anti-MATN2 Rabbit Polyclonal Antibody
Antibodies
Matrilin-2 is part of the Matrilin family of secreted extracelluar matrix proteins that consist of Matrilin -1 to Matrilin-4. Matrilin-2 encodes a member of the von Willebrand factor A domain protein family. It is involved in matrix assembly and development of filamentous networks in the extracellular matrices of various tissues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MATN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty