Anti-BMP-13 Rabbit Polyclonal Antibody

Supplier: BioVision
3823-100
10004-798EA 286.04 USD
10004-798
Anti-BMP-13 Rabbit Polyclonal Antibody
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. These ligands mediate numerous physiological processes. BMPs initiate, promote and maintain cartilage and bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Expression of BMP-13 has been found in the hypertrophic chondrocytes of ossifying long bone centers.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BMP-13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR