Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX1 Rabbit Polyclonal Antibody

Supplier: BioVision

3822-100
10004-794EA 252.35 USD InStock
10004-794
Anti-SOX1 Rabbit Polyclonal Antibody
Antibodies
Sox proteins are related to mammalian sex deternining gene that encodes DNA-binding protein. About 30 SOX family proteins have been identified. Sox-1 is a transcription factor that is found in the embryonic nervous system. Sox-1 maintains nueral cells in an undifferentiated state and has been used as a marker for neural stem cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty