Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MGMT Rabbit Polyclonal Antibody

Supplier: BioVision

3820-100
10004-596EA 252.35 USD InStock
10004-596
Anti-MGMT Rabbit Polyclonal Antibody
Antibodies
MGMT (O-6-methylguanine-DNA methyltransferase) is a DNA repair protein that response to DNA damage by removing methly or alkyl groups from the O6 position of guanine. MGMT protects normal cells from cytotoxic effects and is important in drμg resistance to alkylating agents.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MGMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty