Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EDEM2 Rabbit Polyclonal Antibody

Supplier: BioVision

6633-100
10009-016EA 283.25 USD InStock
10009-016
Anti-EDEM2 Rabbit Polyclonal Antibody
Antibodies
Proteins expressed in the endoplasmic reticulum (ER) are tightly regulated by a variety of quality control mechanisms. Terminally misfolded proteins in the ER are retrotranslocated to the cytoplasm and degraded by proteasomes through a mechanism known as ER-associated degradation (ERAD). EDEM2 (ER degradation-enhancing alpha-mannosidase-like 2) is a 578 amino acid secreted protein that, in conjunction with other EDEM proteins (namely EDEM and EDEM3), is involved in the ERAD pathway of protein degradation. EDEM2, a member of the glycosyl hydrolase 47 family, contains a mannosidase homology domain, an N-terminal cleavable signal sequence and a C-terminal extension that is required for both ER retention and the proper function of EDEM2. Human EDEM2 shares 93% sequence identity with its mouse homolog, suggesting a conserved role between species. Two isoforms of EDEM2 exist due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: EDEM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty