Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT2 Rabbit Polyclonal Antibody

Supplier: BioVision

6632-100
10009-014EA 252.35 USD InStock
10009-014
Anti-SIRT2 Rabbit Polyclonal Antibody
Antibodies
Human sirtuins function as intracellular regulatory proteins. Sirtuin 2 is a member of the class III histone deacetylases (HDACs) and has been implicated in many cellular processes that include histone deacetylation, gene silencing, chromosomal stability, and aging. 

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20-30 µg/ml)

Type: Primary
Antigen: SIRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty