Anti-HDAC3 Rabbit Polyclonal Antibody

Supplier: BioVision
6603-100
10009-010EA 309.39 USD
10009-010
Anti-HDAC3 Rabbit Polyclonal Antibody
HDAC 3 is a member of the class I mammalian histone deacetylase family, which plays an important role in modulating the eukaryotic chromatin structure. Human HDAC 3 is composed of 428 amino acid residues. HDAC 3 interacts with a growing number of transcriptional factors including SMRT and N-CoR. The interacting complexes bind to specific regions of chromatin and regulate gene transcription in these regions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20-30 µg/ml)

Type: Primary
Antigen: HDAC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR