Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody

Supplier: BioVision

6401-05
10008-466EA 231.75 USD InStock
10008-466
Anti-IgG Goat Polyclonal Antibody
Antibodies
Goat anti-rabbit IgG (H+L), horseradish Peroxidase (HPR) conjugated. This antibody has been optimized as secondary antibody in chemiluminescent Western blotting applications.

Recommended Dilutions: Western Blot: 1 µg/ml; a 52 kDa band should be detected

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty