Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FDXR Mouse Monoclonal Antibody [clone: 6C2]

Supplier: BioVision

6216-100
10008-304EA 334.75 USD InStock
10008-304
Anti-FDXR Mouse Monoclonal Antibody [clone: 6C2]
Antibodies
Ferredoxin reductase is a ubiquitous flavoenzyme, containing noncovalently bound FAD as a prosthetic group. It plays a role in delivering NADPH or low potential one-electron donors such as ferredoxin and flavodoxin to redox-based metabolisms in plastids, mitochondria and bacteria. In mammals, ferredoxin reductase is loosely associated with the inner mitochondrial membrane and receives electrons from NADPH. These electrons are transferred to ferredoxin which shuttles electrons to cytochrome P450 in the adrenal cortex mitochondrial steroid hydroxylation systems. It serves as the first electron transfer protein in all the mitochondrial P450 systems. Including cholesterol side chain cleavage in all steroidogenic tissues, steroid 11-beta hydroxylation in the adrenal cortex, 25-OH-vitamin D3-24 hydroxylation in the kidney, and sterol C-27 hydroxylation in the liver.

Recommended Dilutions: WB

Type: Primary
Antigen: Ferredoxin Reductase
Clonality: Monoclonal
Clone: 6C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty