Anti-Proteinase K Rabbit Polyclonal Antibody

Supplier: BioVision
AB286163-100UG
10008-292EA 466.59 USD
10008-292
Anti-Proteinase K Rabbit Polyclonal Antibody
Antibodies
Proteinase K is a serine protease that displays the ability to digest native proteins, thereby inactivating enzymes such as DNase and RNase without recourse to a denaturation process.

Recommended Dilutions: Western Blot: 3 - 6 µg/ml)

0.5 mg/ml polyclonal antibody in phosphate-buffered saline (PBS pH 7.2), containing 30% glycerol, 0.5% BSA, 0.5 mM EDTA, and 0.03% Proclin.

Type: Primary
Antigen: Proteinase K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Tritirachium Album
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR