Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-K2 Rabbit Polyclonal Antibody

Supplier: BioVision

6205-200
10008-286EA 272.95 USD InStock
10008-286
Anti-K2 Rabbit Polyclonal Antibody
Antibodies
Cannabinoids are a class of diverse chemical compounds that activate cannabinoid receptors on cells that repress neurotransmitter release in the brain. They are active chemicals in Cannabis that cause drug-like effects throughout the body, including the central nervous system and the immune system. Anti-K2/spice (synthetic cannabinoids), is a rabbit polyclonal IgG antibody. It has been used in a competitive ELISA format to test the presence of JWH-018, JWH-073, JWH-122, JWH-019, JWH-081, AM-2201 and related compounds and their metabolites in samples such as urine, whole blood, serum, and plasma (see Arntson et al, 2013). Note: If this antibody is used in an immunoassay to detect synthetic cannabinoids, suspect test samples must be confirmed using an alternative analytical method, for example LC-MS-MS.

Recommended Dilutions: ELISA: for 96-well plate coating use 1-3 ?g/mL;

Type: Primary
Antigen: K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty