Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFRA1 Rabbit Polyclonal Antibody

Supplier: BioVision

6201-100
10008-284EA 272.95 USD InStock
10008-284
Anti-GFRA1 Rabbit Polyclonal Antibody
Antibodies
Glial cell line-derived neurotrophic factor (GDNF) is a potent survival factor for central and peripheral neurons and is essential for the development of kidneys and the enteric nerves system. Physiological responses to GDNF require the presence of a novel glycosylphosphadidylinositol linked protein GDNFRα, which is a cell surface receptor for GDNF. The cDNA encoding GDNFRα from human, rat, chicken and mouse have been cloned recently. GDNFRα was also termed Ret ligand 1 (RETL1) or TGF-β-related neurotrophic factor receptor 1 (TrnR1) and nominated as GFRα-1 recently. GFRα-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFRα and the Ret PTK from a complex to transduce GDNF signal and to mediate GDNF function.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: GFRalpha-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty