Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S100 A8 Mouse Monoclonal Antibody [clone: IMG48M7C7]

Supplier: BioVision

6196-100
10008-280EA 406.85 USD InStock
10008-280
Anti-S100 A8 Mouse Monoclonal Antibody [clone: IMG48M7C7]
Antibodies
S100A8 is a low-molecular weight member of the S100 family of calcium-binding protein which promotes tumorigenesis. It promotes cell migration and invasion through p38 MAP dependent NF-kappaB activation leading to an increase of MMP2 and MMP12 in gastric cancer. The phagocyte-specific Ca2+-binding S100A8 protein has been proposed as an essential regulator of the plasma membrane NADPH oxidase activity. It is abundantly expressed in the cytosol of neutrophils and is able to form Ca2+-dependent heterocomplexes, with heterotetramers being a probable prerequisite for its biological activities in myeloid cells. S100A8 and S100A9 have been proposed as essential regulators that exert their role through interactions with NADPH oxidase subunits. S100A8 and S100A9 are generally considered proinflammatory. Whereas hypohalous acids generated by activated phagocytes promote novel modifications in murine S100A8 but modifications to human S100A8 are undefined and there is no evidence that these proteins scavenge oxidants in human disease. Oxidized S100A8 was prominent in lungs from patients with asthma and significantly elevated in sputum compared to controls. Results have broad implications for conditions under which hypohalous acid oxidants are generated by activated phagocytes. Identification in human disease of the novel S100A8 Cys derivatives typical of those generated in vitro strongly supports the notion that S100A8 contributes to antioxidant defense during oxidative stress.

Recommended Dilutions: IHC (P): use antibody at 2 µg/ml, Western blot analysis: 0.5-5 µg/ml.

Type: Primary
Antigen: S100 A8
Clonality: Monoclonal
Clone: IMG48M7C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty