Anti-Thioredoxin Reductase 2 Mouse Monoclonal Antibody [clone: 7B2]

Supplier: BioVision
6165-100
10008-264EA 379.44 USD
10008-264
Anti-Thioredoxin Reductase 2 Mouse Monoclonal Antibody [clone: 7B2]
The mammalian thioredoxin reductases (TrxRs) are a family of seleno-cysteine containing pyridine nucleotide-disulfide oxido-reductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate seleno-cysteine (SeCys) residue in the conserved sequence(-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55 – 58 kDa in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR). TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena. TrxR2 maintains thioredoxin in a reduced state. Implicated in the defenses against oxidative stress. May play a role in redox-regulated cell signaling.

Recommended Dilutions: Western Blot: 1:1000, IP: 1-2 µl.

Type: Primary
Antigen: Thioredoxin Reductase 2
Clonality: Monoclonal
Clone: 7B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR