Anti-CANX Rabbit Polyclonal Antibody

Supplier: BioVision
3811-100
10004-576EA 286.04 USD
10004-576
Anti-CANX Rabbit Polyclonal Antibody
Calnexin (CNX) is a 90kDa integral protein of the endoplasmic reticulum (ER). Calnexin belongs among chaperones, which are characterized by their main function of assisting protein folding and quality control, ensuring that only properly folded and assembled proteins proceed further along the secretary pathway. The function of Calnexin is to retain unfolded or unassembled N-linked glycoproteins in the endoplasmic reticulum.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CANX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR