Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPARG Rabbit Polyclonal Antibody

Supplier: BioVision

3809-100
10004-570EA 252.35 USD InStock
10004-570
Anti-PPARG Rabbit Polyclonal Antibody
Antibodies
PPAR (Peroxisome proliferator-activated receptor) is a member of the nuclear hormone receptor superfamily and functions as a transcriptional activator. PPARg is preferentially expressed in adipocyte as well as in vascular smooth muscle cells and macrophage. Besides its role in mediating adipogenesis and lipids metabolism, PPARg also modulates insulin sensitivity, cell proliferation and inflammation. The inhibition of PPARg transcription activity is due to MAP kinase phosphorylation at serine 84 of PPARg.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PPARG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty