Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HAVCR2 Rabbit Polyclonal Antibody

Supplier: BioVision

3808-100
10004-790EA 252.35 USD InStock
10004-790
Anti-HAVCR2 Rabbit Polyclonal Antibody
Antibodies
As a member of the T cell immunoglobulin and mucin domain (TIM) family, Tim-3 is reported to promote or terminate Th1 immunity and is thought to influence a range of inflammatory conditions. It is constitutively expressed in a subset of murine IFNgamma-secreting Th1 cells, but not Th2 cells, in both human and mice. Additionally, Tim-3 is expressed dendritic cells, and by a subset of CD4+ cells where its levels of expression increase upon anti-CD3/28 stimulation. This molecule is also highly expressed in in vitro polarized Th1 cells, and at lower levels in Th17 cells. It is known to synergize with Toll-like receptors and is thought to regulate Th1 and Th17 cytokine secretion.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: Tim-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty