Anti-LMNB1 Rabbit Polyclonal Antibody

Supplier: BioVision
3807-100
10004-786EA 286.04 USD
10004-786
Anti-LMNB1 Rabbit Polyclonal Antibody
Catalase is known marker for peroxisomes. It is the most abundant protein in the peroxisomes. It is present in all aerobically respiring organisms. It protects cells form the toxicity of hydrogen peroxide.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LMNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR