Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAT Rabbit Polyclonal Antibody

Supplier: BioVision

3806-100
10004-782EA 252.35 USD InStock
10004-782
Anti-CAT Rabbit Polyclonal Antibody
Antibodies
Catalase is known marker for peroxisomes. It is the most abundant protein in the peroxisomes. It is present in all aerobically respiring organisms. It protects cells form the toxicity of hydrogen peroxide.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty