Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Prohibitin Rabbit Polyclonal Antibody

Supplier: BioVision

3805-100
10004-778EA 252.35 USD InStock
10004-778
Anti-Prohibitin Rabbit Polyclonal Antibody
Antibodies
Prohibitin is an evolutionarily conserved protein that has anti-proliferative activity. Prohibitin is a 30 kDa, post-synthetically modified protein found in the inner membrane of mitochondria and on the plasma membrane of B cells. It mediates B cell mutation. Prohibitin Controls cell cycle in both transcriptional and post-translational mechanisms. Prohibitin is a potential tumor suppressor. Prohibitin gene mutations are correlated with the breast cancer development and/or progression in over 80%0f the cell lines analyzed.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; Iimmunofluorescence: 5-10 µg/ml

Type: Primary
Antigen: Prohibitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty