Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DKK2 Rabbit Polyclonal Antibody

Supplier: BioVision

3802R-100
10004-776EA 252.35 USD InStock
10004-776
Anti-DKK2 Rabbit Polyclonal Antibody
Antibodies
Xenopus Dickkopf (Dkk)-1 was initially discovered as a Wnt antagonist that plays an important role in head formation. By far, four members of Dkk have been identified in mammals. Each Dkk molecule contains two conserved cysteine-rich domains. Recent studies showed that the second Cys-rich domains of Dkk1 and Dkk2 inhibited Wnt-3a-activated signaling, whereas the first Cys-rich domains had no effects. In addition, the second Cys-rich domain of Dkk-2, but not that of Dkk-1, was able to activate the canonical pathway in the presence of LRP6, and this LRP-dependent signaling does not require Dvl.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty