Anti-DKK2 Rabbit Polyclonal Antibody

Supplier: BioVision
3802A-100
10004-772EA 286.04 USD
10004-772
Anti-DKK2 Rabbit Polyclonal Antibody
Xenopus Dickkopf (Dkk)-1 was initially discovered as a Wnt antagonist that plays an important role in head formation. By far, four members of Dkk have been identified in mammals. Each Dkk molecule contains two conserved cysteine-rich domains. Recent studies showed that the second Cys-rich domains of Dkk1 and Dkk2 inhibited Wnt-3a-activated signaling, whereas the first Cys-rich domains had no effects. In addition, the second Cys-rich domain of Dkk-2, but not that of Dkk-1, was able to activate the canonical pathway in the presence of LRP6, and this LRP-dependent signaling does not require Dvl.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR