Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MEN1 Rabbit Polyclonal Antibody

Supplier: BioVision

3797-100
10005-926EA 252.35 USD InStock
10005-926
Anti-MEN1 Rabbit Polyclonal Antibody
Antibodies
Menin (multiple endocrine neoplasia I, MEN1, MEAI, SCG2) is a nuclear tumor suppressor that is mutated in patients with multiple endocrine neoplasia type I (MEN1). Menin can activate the transcription of differentiation-regulating genes by covalent histone modification. In osteoblasts, the interaction of menin and the TGFβ/Smad3 pathway negatively regulates BMP2/Smad1/5- and Runx2-dependent transcription activities leading to inhibition of late-stage differentiation. Menin regulates the expression of IGFBP-2 by influencing the IGFBP-2 promoter. Ectopic overexpression of menin via adenoviruses induces apoptosis in murine embryonic fibroblasts in a Bax/Bak-dependent manner. Two mRNA exist and two variants of the shorter mRNA have alternative splicing that changes the CDS. Five variants where alternative splicing takes place in the 5' UTR have been identified.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MEN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty