Anti-RBBP5 Rabbit Polyclonal Antibody

Supplier: BioVision
3793R-100
10005-920EA 286.04 USD
10005-920
Anti-RBBP5 Rabbit Polyclonal Antibody
Retinoblastoma binding protein 5 (RbBP5) belongs to a highly conserved subfamily of WD-repeat proteins and it is also a subunit of a human Set1-like histone methyltransferase (HMT) complex. The encoded RbBP 5protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: RBBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR