Anti-RBBP5 Rabbit Polyclonal Antibody

Supplier: BioVision
3793-100
10005-918EA 402.79 USD
10005-918
Anti-RBBP5 Rabbit Polyclonal Antibody
Retinoblastoma binding protein 5 (RbBP5) is a subunit of a human Set1-like histone methyltransferase (HMT) complex. The Set-1 like complexes are responsible for mono-, di-, and tri-methylation of histone H3 K4. RbBP5 associates with a multitude of proteins in a number of Set1-like complexes which include various combinations of MLL, MLL2, MLL3, MLL4, ASH2L,WDR5, hDPY-30, hSwd2, CXXC1, HCF1, Menin, PTIP, PA1, NCOA6, and UTX. RbBP5 is also one of many proteins that bind the Retinoblastoma tumor suppressor protein, pRb.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunoprecipitation

Type: Primary
Antigen: RBBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR