Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRMT1 Rabbit Polyclonal Antibody

Supplier: BioVision

3792-100
10005-916EA 252.35 USD InStock
10005-916
Anti-PRMT1 Rabbit Polyclonal Antibody
Antibodies
PRMT1 methylate’s (mono & asymmetric dimethylation) the guanidino nitrogens of arginyl residues present in a glycine and arginine-rich domain (may methylate HNRNPA1 and histones) methylate’s SUPT5H. The PRMT1 protein functions as a histone methyltransferase specific for H4. PRMT1 is an essential factor in oncogenesis and is a potential novel therapeutic target in cancer. PRMT1-mediated methylation serves as a positive modulator of IR/IRS-1/PI3K pathway and glucose uptake in skeletal muscle cells. CAF1 is a new regulator of PRMT1-dependent arginine methylation. PRMT1 arginine-methylate’s MRE11 therefore it regulates the activity of MRE11-RAD50-NBS1 complex during the intra-S-phase DNA damage checkpoint response. PRMT1 plays a post-translationally part in regulating the transcriptional activity. PRMT1 is found predominantly in the cytoplasm, though a fraction of PRMT1 is located in the nucleus.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: PRMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty