Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Rho Kinase Alpha Rabbit Polyclonal Antibody

Supplier: BioVision

3789-100
10004-766EA 252.35 USD InStock
10004-766
Anti-Rho Kinase Alpha Rabbit Polyclonal Antibody
Antibodies
Rho-associated protein kinase 2 (ROCK2), or Rho kinase alpha, is a serine-threonine protein kinase that plays an important role in cellular apoptosis, growth, cell motility and cell contraction. ROCK2 shares an overall 65% homology in amino acid sequence with ROCK1. The ROCK1 and ROCK2 isoforms are products of two separate genes. Both ROCK1 and ROCK2 are downstream targets of the Rho A small GTP-binding protein. As downstream effectors of RhoA, ROCK1 and ROCK2 mediate changes in the actin cytoskeleton via phosphorylation of target proteins such as the MBS subunit of MLCP, MLC, ERM proteins, protein LIM kinase and adducin.

Recommended Dilutions: Western Blot: 1:1000-1:2500; Immunoprecipitation

Type: Primary
Antigen: Rho Kinase Alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty