Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIM1 Rabbit Polyclonal Antibody

Supplier: BioVision

3787R-100
10004-762EA 252.35 USD InStock
10004-762
Anti-PIM1 Rabbit Polyclonal Antibody
Antibodies
The proto-oncogene Pim1 belongs to a family of serine/threonine protein kinases that are highly conserved throμgh evolution in multicellular organisms. Pim1 is involved in the control of cytokine-mediated cell proliferation, differentiation and survival of lymphoid and myeloid cells as well as others. Expression of Pim1 can be stimulated by a variety of growth factors and is regulated at four different levels: transcriptional, post-transcriptional, translational and post-translational. Recently, Pim1 has been shown to enhance the activities of p100, c-Myb and Cdc 25a and in part this might explain reported effects of Pim1 on mitogenesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty