Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LOK Rabbit Polyclonal Antibody

Supplier: BioVision

3785-100
10004-758EA 252.35 USD InStock
10004-758
Anti-LOK Rabbit Polyclonal Antibody
Antibodies
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. LOK (Lymphocyte-oriented kinase), also known as STK10 (serine/threonine kinase 10), is a 968 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Expressed in lymphoid organs, LOK functions to catalyze the ATP-dependent phosphorylation of target proteins, such as MBP (myelin basic protein) and Histone H2A, thereby playing a role in signaling pathways throughout the cell.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunoprecipitation

Type: Primary
Antigen: LOK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty