Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JIK Rabbit Polyclonal Antibody

Supplier: BioVision

3784-100
10004-756EA 252.35 USD InStock
10004-756
Anti-JIK Rabbit Polyclonal Antibody
Antibodies
JNK/SAPK-inhibitory kinase (JIK) is a serine/threonine kinase that belongs to the STE20 kinase family. The kinase domain of JIK is similar to the GCK-like subfamily of STE20 kinases, while its non-catalytic domain is similar to a Caenorhabditis elegans putative serine/threonine kinase, SULU. JIK inhibits c-Jun NH2-terminal kinase/stress-activated protein kinase (JNK/SAPK), which is activated by many types of cellular stresses and extracellular signals. JNK/SAPK regulates cell survival, apoptosis and proliferation in mouse development. JIK is negatively regulated by epidermal growth factor (EGF) and tyrosine kinase receptors. In unstimulated human T cells, JIK is cytoplasmic, whereas in the continuously dividing human T cells of Jurkat lymphoma, JIK is nuclear.

Recommended Dilutions: Western Blot: 1:500-1:2500; Immunoprecipitation

Type: Primary
Antigen: JIK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty