Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TPR Rabbit Polyclonal Antibody

Supplier: BioVision

3781-100
10004-750EA 252.35 USD InStock
10004-750
Anti-TPR Rabbit Polyclonal Antibody
Antibodies
The vertebrate nuclear pore complex (NPC) is a macromolecular assembly of protein subcomplexes forming a structure of eightfold radial symmetry. The NPC core consists of globular subunits sandwiched between two coaxial ring-like structures of which the ring facing the nuclear interior is capped by a fibrous structure called the nuclear basket. The assembly of the NPC is a stepwise process in which Trp-containing peripheral structures assemble after other components, including p62. TPR localizes to intranuclear filaments of the NPC, and is a component of the cytoplasmic fibrils of the NPC implicated in nuclear protein import. Experimental data suggest that TPR is tethered to intranuclear filaments of the NPC by its coiled coil domain leaving the acidic COOH terminus free to interact with soluble transport factors and mediate export of macromolecules from the nucleus.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: TPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty