Anti-RCH1 Rabbit Polyclonal Antibody

Supplier: BioVision
3779-100
10004-746EA 402.79 USD
10004-746
Anti-RCH1 Rabbit Polyclonal Antibody
The import of proteins from the cytoplasm to the nucleus involves docking the protein to receptors associated with the nuclear pore complex followed by translocation through the nuclear pore. RCH1 is an adaptor protein that recognizes the nuclear localization signal (NLS) on cargo and binds the karyopherin importin-beta receptor for nuclear import which is regulated by RanGTP.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunoprecipitation

Type: Primary
Antigen: RCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR