Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSE1 Rabbit Polyclonal Antibody

Supplier: BioVision

3778-100
10004-744EA 252.35 USD InStock
10004-744
Anti-CSE1 Rabbit Polyclonal Antibody
Antibodies
CSE1 function to re-export importin-alpha to the cytoplasm after it has transported nuclear localization signal (NLS) containing proteins to the nucleus. CSE1 is also thought to play a role in cell proliferation and the mitotic spindle checkpoint.

Recommended Dilutions: Western Blot: 1:1000-1:2500; Immunoprecipitation

Type: Primary
Antigen: CSE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty