Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAPDH Rabbit Polyclonal Antibody

Supplier: BioVision

3777R-100
10004-564EA 278.65 USD InStock
10004-564
Anti-GAPDH Rabbit Polyclonal Antibody
Antibodies
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), also called uracil DNA glycosylase, catalyzes the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD), an important energy-yielding step in carbohydrate metabolism. While GAPDH has long been recognized as playing an integral role in glycolysis, additional functions of GAPDH include acting as a uricil DNA glycosylase, activating transcription, binding RNA and involvement in nuclear RNA export, DNA replication and DNA repair. Expression of GAPDH is upregulated in liver, lung and prostate cancers. GAPDH translocates to the nucleus during apoptosis. GAPDH complexes with neuronal proteins implicated in human neuro-degenerative disorders including the b-Amyloid precursor, Huntingtin and other triplet repeat neuronal disorder proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: GAPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty