Anti-TAOK1 Rabbit Polyclonal Antibody

Supplier: BioVision
3776R-100
10004-560EA 285.95 USD
10004-560
Anti-TAOK1 Rabbit Polyclonal Antibody
Serine/threonine-protein kinase TAO1 is an enzyme that in humans is encoded by the TAOK1 gene.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TAOK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR