Anti-PRDX4 Mouse Monoclonal Antibody [clone: 3A1]

Supplier: BioVision
6161-100
10008-256EA 379.44 USD
10008-256
Anti-PRDX4 Mouse Monoclonal Antibody [clone: 3A1]
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, particularly amino-terminal region containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20 – 30 kDa in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol. Prx IV is probably involved in redox regulation of the cell. Regulates the activation of NFkappaB in the cytosol by a modulation of I-kappa-B-alpha phosphorylation.

Recommended Dilutions: Western Blot: 2-4 µl/ml, IP: 1-2 µl, ICC: 20 µl/ml.

Type: Primary
Antigen: PRDX4
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR