Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TDG Rabbit Polyclonal Antibody

Supplier: BioVision

6157-100
10008-248EA 345.05 USD InStock
10008-248
Anti-TDG Rabbit Polyclonal Antibody
Antibodies
In the DNA of higher eukaryotes, hydrolytic deamination of 5­methylcytosine to thymine leads to the formation of G/T mismatches. G/T mismatch specific Thymine DNA Glycosylase (TDG) is a nuclear protein which corrects G/T mismatches to G/C pairs by hydrolyzing the carbon nitrogen bond between the sugar phosphate backbone of the DNA and the mispaired thymine. TDG also corrects a subset of G/U mispairs inefficiently removed by the more abundant uracil glycosylases. Retinoic acid receptors interact physically and functionally with TDG, enhancing the ability of the retinoid X receptor and the retinoid X receptor/retinoid acid receptor complex to bind to their response elements. TDG interacts with, and is covalently modified by, the ubiquitin­like proteins SUMO­1 and SUMO­2/3, resulting in a reduction of the DNA substrate and AP site binding affinity of TDG. This sumoylation is associated with a significant increase in enzymatic turnover in reactions with a G/U substrate and the loss of G/T processing activity.

Recommended Dilutions: Western Blot: 1:500-1:1000.

Type: Primary
Antigen: TDG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty