Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SETDB1 Rabbit Polyclonal Antibody

Supplier: BioVision

6155-100
10008-244EA 345.05 USD InStock
10008-244
Anti-SETDB1 Rabbit Polyclonal Antibody
Antibodies
The Erg­associated protein with SET domain (ESET), also known as SET domain, bifurcated 1 (SETDB1) protein, is a member of a family of histone lysine methyltransferases, each of which contains a conserved catalytic SET domain originally identified in Drosophila, Enhancer of zeste, and Trithorax proteins. ESET also contains tudor and methyl­CpG­binding domains, which may coordinate binding to methylated histones and methylated DNA, respectively. ESET methylates histone H3 Lys9, creating a transcriptionally repressive mark that facilitates gene silencing. However, unlike SUV39H histone H3 Lys9 methyltransferases, which function mainly in heterochromatin regions such as pericentric heterochromatin, ESET functions mainly in euchromatic regions to repress gene promoters. ESET interacts with a variety of proteins, including transcription factors (ERG), histone deacetylases (HDAC1/2), DNA methyltransferases (DNMT3A/B) and transcriptional co­repressors (mSin3A/B, MBD1, KAP­1, the ATFa­associated modulator mAM). mAM forms a complex with ESET, stimulating its methyltransferase activity, specifically the conversion of di­methyl to tri­methyl histone H3 Lys9. MBD1 recruits ESET to the CAF­1 complex to facilitate methylation of histone H3 Lys9 during replication­coupled chromatin assembly in S phase. DNMT3A recruits ESET to silenced promoters in cancer cells. ESET may play a role in the pathogenesis of Huntington's disease, since levels of ESET protein and tri­methyl histone H3 Lys9 are both increased in diseased brains.

Recommended Dilutions: Western Blot: 1:500-1:1000, IHC: 1:50-1:100, IP: 1:50-1:100

Type: Primary
Antigen: SETDB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty