Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NCOA1 Rabbit Polyclonal Antibody

Supplier: BioVision

6153-100
10008-240EA 345.05 USD InStock
10008-240
Anti-NCOA1 Rabbit Polyclonal Antibody
Antibodies
There are three members of the steroid receptor co­activator (SRC) family of proteins: SRC­1 (NCoA­1), SRC­2 (TIF2/GRIP1/NCoA­2), and SRC­3 (ACTR /TRAM­1/AIB1). All SRC family members share significant structural homology and function to stimulate transcription mediated by nuclear hormone receptors and other transcriptional activators such as Stat3, NF­κB, E2F1, and p53. Two SRC proteins, SRC­1 and SRC­3, function as histone acetyltransferases. In addition, all three family members can recruit other histone acetyltransferases (CBP/p300, PCAF) and histone methyltransferases (PRMT1, CARM1) to target promoters and cooperate to enhance expression of many genes. The SRC proteins play important roles in multiple physiological processes including cell proliferation, cell survival, somatic cell growth, mammary gland development, female reproductive function, and vasoprotection. SRC­1 and SRC­3 are conduits for kinase mediated growth factor signaling to the estrogen receptor and other transcriptional activators. Seven SRC­1 phosphorylation sites and six SRC­3 phosphorylation sites have been identified, which are induced by steroids, cytokines, and growth factors and involve multiple kinase signaling pathways. Research has shown that all three SRC family members are associated with increased activity of nuclear receptors in breast, prostate, and ovarian carcinomas. According to the literature, SRC­3 is frequently amplified or overexpressed in a number of cancers, and SRC­1/PAX3 and SRC­2/MYST3 translocations are found associated with rhabdomyosarcoma and acute myeloid leukemia, respectively.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200.

Type: Primary
Antigen: NCOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty