Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EHMT2 Rabbit Polyclonal Antibody

Supplier: BioVision

6152-100
10008-238EA 345.05 USD InStock
10008-238
Anti-EHMT2 Rabbit Polyclonal Antibody
Antibodies
G9a, also known as Euchromatic histone­lysine N­methyltransferase 2 (EHMT2), is a member of a family of histone lysine methyltransferases. Recombinant G9a can mono­, di­ and tri­methylate histone H3 on Lys9 and Lys27 in vitro. However, in vivo G9a forms a complex with GLP, a G9a­related histone methyltransferase, and together these proteins function as the major euchromatic histone H3 Lys9 mono­ and di­methyltransferases, creating transcriptionally repressive marks that facilitate gene silencing. G9a methylates itself on Lys165, a modification that regulates the association of HP1 repressor proteins with the G9a/GLP complex. The G9a/GLP complex also contains Wiz, a zinc finger protein that is required for G9a/GLP hetero­dimerization and complex stability. Wiz contains two CtBP co­repressor binding sites, which mediate the association of the G9a/GLP with the CtBP co­repressor complex. In addition, G9a and GLP are components of other large transcriptional co­repressor complexes, such as those involving E2F6 and CDP/cut. G9a interacts with DNMT1, and both proteins are required for methylation of DNA and histone H3 (Lys9) at replication foci, providing a functional link between histone H3 Lys9 and CpG methylation during DNA replication. G9a activity is critical for meiotic prophase progression, as mutant mice deficient in germ line G9a show a large loss of mature gametes. In addition, G9a facilitates increased global levels of di­methyl histone H3 (Lys9) during hypoxic stress and increased G9a expression is associated with hepatocellular carcinoma.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200.

Type: Primary
Antigen: EHMT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty