Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMI 1 Rabbit Polyclonal Antibody

Supplier: BioVision

6150-100
10008-234EA 345.05 USD InStock
10008-234
Anti-BMI 1 Rabbit Polyclonal Antibody
Antibodies
The polycomb group (PcG) of proteins contributes to the maintenance of cell identity, stem cell self-­renewal, cell cycle regulation, and oncogenesis by maintaining the silenced state of genes that promote cell lineage specification, cell death, and cell cycle arrest. PcG proteins exist in two complexes that cooperate to maintain long term gene silencing through epigenetic chromatin modifications. The first complex, EED­EZH2, is recruited to genes by DNA­binding transcription factors and methylates histone H3 on Lys27. This histone methyl­transferase activity requires the Ezh2, Eed, and Suz12 subunits of the complex. Histone H3 methylation at Lys27 facilitates the recruitment of the second complex, PRC1, which ubiquitinylates histone H2A on Lys119. Bmi1 is a component of the PRC1 complex, which together with Ring1 strongly enhances the E3 ubiquitin ligase activity of the Ring2 catalytic subunit. Bmi1 plays an important role in the regulation of cell proliferation and senescence through repression of the p16 INK4A and p19 ARF genes and is required for maintenance of adult hematopoietic and neural stem cells.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200.

Type: Primary
Antigen: BMI 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty